missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Aluminum Cups for Pellet Press PP 25

Catalog No. p-4701900
Change view
Click to view available options
Diameter (Metric):
32 mm
40 mm
Includes:
1000 pieces
600 pieces
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Includes
08418519 32 mm 1000 pieces
08418520 40 mm 600 pieces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08418519 Supplier RETSCH Supplier No. 22.005.0001
Only null left

Specifications

Type Aluminum Cup
Diameter (Metric) 32 mm
For Use With (Equipment) Pellet Press PP 25
Includes 1000 pieces
Material Aluminum