Learn More
Kent Scientific Retractor Wire for SurgiSuite Multi-Functional Surgical Platforms
Catalog No. 13005188
Encompass
Change view
Click to view available options
Length (Metric):
10 cm
14 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
13005188 14 cm
13005187 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13005188 Supplier Kent Scientific Supplier No. SURGI5013
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with SurgiSuite Multi-Functional Surgical Platforms. Kent Scientific Retractor Wire engage and lock into fixator jaws at desired elevation.
  • Engage and lock into fixator jaws at desired elevation
Specifications
For Use With (Application) Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction
Type Retractor Wire
For Use With (Equipment) SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms
Length (Metric) 14 cm