Learn More
Kent Scientific Retractor Tip for SurgiSuite Multi-Functional Surgical Platforms
Catalog No. 13005189
Encompass
Change view
Click to view available options
Size (Metric):
1 mm
5 mm
7.5 mm
Tip Style:
Blunt
Sharp
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Tip Style
13005189 1 mm Sharp
13005190 1 mm Blunt
13005192 5 mm Blunt
13005193 7.5 mm Blunt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13005189 Supplier Kent Scientific Supplier No. SURGI5014
For use with SurgiSuite Multi-Functional Surgical Platforms. Kent Scientific Retractor Tip is locked onto wire for static retraction, or affixed to wire for dynamic retraction.
  • Locked onto wire for static retraction
  • Affixed to elastic wire for dynamic retractor
Specifications
For Use With (Application) Surgery, Homeothermic Warming, Surgical Procedures, Tissue Retraction
Type Retractor Tip
For Use With (Equipment) SurgiSuite and SurgiSuite-LG Multi-Functional Surgical Platforms
Size (Metric) 1 mm
Tip Style Sharp