Learn More
Fine Science Tools RETRACTOR BLUNT 1MM WIDE
Catalog No. NC9164748
Encompass
Product: Magnetic Fixator Retraction System - Blunt Retractors/1mm Wide/ Item: Kit for Animals up to 200g/ Maximum Spread: Adjustable/ Unit: 7 Piece Kit
Catalog No. NC9164748 Supplier Fine Science Tools Supplier No. 1820009
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product: Magnetic Fixator Retraction System - Blunt Retractors/1mm Wide/ Item: Kit for Animals up to 200g/ Maximum Spread: Adjustable/ Unit: 7 Piece Kit