missing translation for 'onlineSavingsMsg'
Learn More

Retinoic Acid 93.0+%, TCI America™

Catalog No. p-7135678
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00641G 1 g
R00645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00641G Supplier TCI America Supplier No. R00641G

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
MDL Number MFCD00001551
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
Synonym retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra
PubChem CID 444795
ChEBI CHEBI:15367
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O

Specifications

Melting Point 180°C
Color Yellow
Quantity 1 g
Formula Weight 300.44
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Retinoic Acid
RTECSNumber VH6475000
TSCA Yes