Learn More
StatLab Reticulum Fibers Control Slides
Catalog No. 22024936
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
22024936 25 Pk.
1 options
Catalog No. 22024936 Supplier StatLab Supplier No. CSR0125P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use reticulum fibers control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 25 Pk.