Learn More
Synthware™ Replacement Plug Retention Kit
GREENER_CHOICE
Catalog No. 31502040
Change view
Click to view available options
Plug Bore:
12/30 mm
16.2/56 mm
17.35/50 mm
17/40 mm
20/44 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31502040 20/44 mm
31502036 12/30 mm
31502037 17/40 mm
31502038 17.35/50 mm
31502039 16.2/56 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31502040 Supplier Synthware™ Supplier No. S692044
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with stopcocks
Specifications
Type Replacement Stainless Steel Retention Kit
Plug Bore 20/44 mm
Material Stainless Steel
For Use With (Application) Organic Synthesis
For Use With (Equipment) Glass Stopcock