Learn More
SignalChem Biotech RET (L790F), Active 50ug
Catalog No. 502573423
Encompass
Recombinant human RET (L790F) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573423 Supplier SignalChem Biotech Supplier No. R0212LG50
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human RET (L790F) (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.