Learn More
Restek™ Universal Press-Tight™ Connectors: Universal Connector
Connect analytical columns, guard columns, transfer lines or detectors
Supplier: Restek™ RK20400
Description
- Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
- Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
- Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
| Connector | |
| 5 Pack |