Learn More
Restek™ Universal 10-32 PEEK Column Connector
Universal PEEK connectors allow easy installation of all 0.0625 in. tubing, including stainless steel, up to 5000psi.
Supplier: Restek™ RK25016
Description
- Quantity: 10/Pk.
Specifications
| Connector | |
| Black | |
| 10/Pk. | |
| Universal 10-32 PEEK Column Plug |