Learn More
Restek™ Solvent Debubbler
Removes bubbles from the fluid stream before it enters the pump in an LC system.
Supplier: Restek™ RK 25014
Description
Uses a small reservoir to remove bubbles from the inlet fluid stream to prevent check valve malfunctions and pump cavitation. Unique design allows bubbles in the inlet stream to rise and be trapped in the reservoir. Gas/liquid interface is easily visible through the translucent wall of the device. Loosening the airtight cap releases the trapped gas.
- Polypropylene body and bracket
- Universal inlet and outlet connecting tips are made of 316 stainless steel and accept 0.063 in., 0.085 in., and 0.125 in. ID tubing
Specifications
| Solvent Debubbler | |
| Bracket | |
| Polypropylene |