Learn More
Restek™ Soap Film Bubble Flowmeter
For flows between 0.1 and 10 cc/min. requires 1mL flowmeter
$221.21 - $282.15
Specifications
Product Type Bubble Flowmeter
View More Specs
Includes: A reservoir bulb, 24 in. (60cm) of 1/4 in. ID tubing, adapter tubes for 1/8 in. tubing and 0.53mm ID capillary columns, and Velcro™ fasteners.
Products 2
Catalog Number Mfr. No. Flow Rate Volume (English) Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Volume (English) Volume (Metric) Price Quantity & Availability  
06710511
Encompass
View Documents Promotion Details
Restek™
RK 20135
0.1 and 10 cc/min. 0.03 oz. 1 mL
Each for $221.21
Only null left
06710512
Encompass
View Documents Promotion Details
Restek™
RK 20136
10 and 300 cc/min. 1.69 oz. 50 mL
Each for $282.15
Only null left
Description
Description
  • 50mL flowmeter for flows between 10 and 300cc/min.
Specifications
Specifications
Bubble Flowmeter