Learn More
Restek™ Septum Nuts for use with Agilent GCs
For use with Agilent 5890, 6850, 6890, 7890 split/splitless injectors.
$196.55 - $215.70
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
06710861
|
Restek™
RK 20631 |
Manual Injection (for 26-Gauge Needles) |
Each for $196.55
|
|
|||||
06711512
|
Restek™
RK 21309 |
Autosampler and PTV (for 23-Gauge Needles) |
Each for $215.70
|
|
|||||
Description
- Needle guide allows easy penetration and prevents septum coring
- Made of high-quality stainless steel
- Manual injection septum nut allows use of 26-gauge needles for on-column injections
- Autosampler and PTV septum nut allows use of 23-gauge needles
Specifications
| Septum Nut |