Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Catalog No. 06712019
Encompass
Change view
Click to view available options
Product Type:
Shell Weldmen
Shell Weldment
Weldment
Weldment with EPC
Material:
Siltek Treated Stainless Steel
Stainless Steel
For Use With (Equipment):
Agilent 5890 GCs, Agilent 6890/6850 GCs
Agilent 6890/6850 GCs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material For Use With (Equipment)
06712019 Weldment with EPC Stainless Steel Agilent 6890/6850 GCs
06710552 Weldment Stainless Steel Agilent 5890 GCs, Agilent 6890/6850 GCs
06712018 Shell Weldmen Stainless Steel Agilent 6890/6850 GCs
06712016 Shell Weldment Siltek Treated Stainless Steel Agilent 6890/6850 GCs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06712019 Supplier Restek™ Supplier No. RK 22674
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement weldments for Agilent 5890 and 6890/6850 GCs
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent 6890/6850 GCs
Material Stainless Steel
Product Type Weldment with EPC