Learn More
Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments
Replacement weldments for Agilent 5890 and 6890/6850 GCs
Supplier:  Restek™ RK 22673
Encompass
 View more versions of this product
Catalog No. 06712018
Only null left
Description
Description
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
Chemical Identifiers
Gas Chromatography
Specifications
Gas Chromatography
Agilent 6890/6850 GCs
Stainless Steel
Shell Weldmen
Product Suggestions
Product Suggestions