Learn More
Restek™ Replacement Dual Vespel™ Ring Inlet Seals
Catalog No. 06802809
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.2 mm
Material:
Gold-plated
Siltek
Stainless Steel
Packaging Quantity:
10/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Material Packaging Quantity
06802809 1.2 mm Gold-plated 10/Pk.
06802807 1.2 mm Stainless Steel 10/Pk.
06802805 0.8 mm Siltek 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06802809 Supplier Restek™ Supplier No. RK 21247
Only null left
Replacement seals for Agilent™ 5890/6890/6850 split/splitless injection ports
  • Increase column lifetime because oxygen cannot permeate into the carrier gas
  • Reduced noise benefits high-sensitivity detectors (ECDs or MSDs)
  • Special grade of stainless steel that is softer and deforms more easily, creating a better seal
  • Gold-plated seals offer better inertness than standard stainless steel
  • Siltek™ seals feature inertness similar to that of fused silica capillary columns
  • All seals include washers
  • 1.2mm inlet seal is recommended for use with Vespel™/graphite ferrules or when installing two columns using a two-hole ferrule
  • 0.8mm inlet seal is recommended for use with graphite ferrules and single capillary column installation
Specifications
Type Seal
Diameter (Metric) Inner 1.2 mm
Packaging Quantity 10/Pk.
Material Gold-plated