Learn More
Restek™ Replacement Dual Vespel™ Ring Inlet Seals DFS Item
Replacement seals for Agilent™ 5890/6890/6850 split/splitless injection ports
Supplier:  Restek™ RK21238
 View more versions of this product
Catalog No. 06-802-800
Only null left
Explore available promotions
Description
Description
  • Increase column lifetime because oxygen cannot permeate into the carrier gas
  • Reduced noise benefits high-sensitivity detectors (ECDs or MSDs)
  • Special grade of stainless steel that is softer and deforms more easily, creating a better seal
  • Gold-plated seals offer better inertness than standard stainless steel
  • Siltek™ seals feature inertness similar to that of fused silica capillary columns
  • All seals include washers
  • 1.2mm inlet seal is recommended for use with Vespel™/graphite ferrules or when installing two columns using a two-hole ferrule
  • 0.8mm inlet seal is recommended for use with graphite ferrules and single capillary column installation
Specifications
Specifications
Seal
0.8 mm
2/Pk.
Stainless Steel
Product Suggestions
Product Suggestions