missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Q-sep™ QuEChERS dSPE Tubes for Extract Clean-Up

Catalog No. 06729303
Encompass
Change view
Click to view available options
Volume (Metric):
2 mL
15 mL
Sorbent:
Ultra pure, PSA
Ultra pure, PSA, C18
Ultra pure, PSA, GCB
Material:
150mg MgSO4
900mg MgSO4
Shake Time:
0.5 min.
2 min.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sorbent Material Shake Time
06729303 15 mL Ultra pure, PSA, GCB 900mg MgSO4 2 min.
06729300 2 mL Ultra pure, PSA, GCB 150mg MgSO4 2 min.
06729301 2 mL Ultra pure, PSA 150mg MgSO4 0.5 min.
06729302 2 mL Ultra pure, PSA, C18 150mg MgSO4 0.5 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06729303 Supplier Restek™ Supplier No. RK 26126

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fast, simple sample prep for multiresidue pesticide analysis.

QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep products make QuEChERS procedures even simpler. All extraction salts, sorbents, and sample tubes are included. Multiple sorbents are used to extract different types of interferences.

  • Ready-to-use tubes, no glassware required
  • Preweighed, ultra-pure sorbents
  • Packaged in foil subpacks of 10 for enhanced protection and storage stability
  • Support original unbuffered, AOAC (2007.01), European (EN 15662), and mini-multiresidue QuEChERS methods

Specifications

For Use With (Application) Fast, Simple Sample Prep for Multi Residue Pesticide Analysis
Sorbent Ultra pure, PSA, GCB
Type Q-sep QuEChERS dSPE Micro-centrifuge Tube
Volume (Metric) 15 mL
Material 900mg MgSO4
Method AOAC 2007.01
Sample Volume 6 mL
Shake Time 2 min.