Learn More
Restek™ Q-sep™ QuEChERS dSPE Tubes for Extract Clean-Up
Fast, simple sample prep for multiresidue pesticide analysis.
Supplier: Restek™ RK26124
Description
QuEChERS methods are fast, easy, and cost-effective, and Restek Q-sep products make QuEChERS procedures even simpler. All extraction salts, sorbents, and sample tubes are included. Multiple sorbents are used to extract different types of interferences.
- Ready-to-use tubes, no glassware required
- Preweighed, ultra-pure sorbents
- Packaged in foil subpacks of 10 for enhanced protection and storage stability
- Support original unbuffered, AOAC (2007.01), European (EN 15662), and mini-multiresidue QuEChERS methods
Specifications
| Fast, Simple Sample Prep for Multi Residue Pesticide Analysis | |
| Q-sep QuEChERS dSPE Micro-centrifuge Tube | |
| 150mg MgSO4 | |
| 1 mL |
| Ultra pure, PSA | |
| 2 mL | |
| AOAC 2007.01 | |
| 0.5 min. |