Learn More
Restek™ Purge-and-Trap Spargers for Tekmar 2000, 3000 or 3100 DFS Item
For use with Tekmar purge-and-trap instruments only.
$320.28 - $320.28
Products 1
Catalog Number Mfr. No. Type Capacity Price Quantity  
Catalog Number Mfr. No. Type Capacity Price Quantity  
06711454 DFS Item
View Documents
Restek™
RK21151
Fritted 25 mL
Each for $320.28
Only null left
Description
Description
  • Available with uniform frits to ensure maximum purging efficiency
  • Manufactured to tight tolerances to ensure a leak-tight seal
  • Fritted, 0.5 in. mount
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'