Learn More
Restek™ Packed Column Inlet Adapter Kit for 1/4-Inch Injection Ports
Centers column perfectly in injection port to eliminate bent syringe needles.
$149.66 - $157.08
| Catalog Number | Mfr. No. | For Use With | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With | Price | Quantity & Availability | |||||
06711705
|
Restek™
RK 21651 |
0.25 in. Injection Port, 0.25 to 0.125 in. Column |
Each for $149.66
|
|
|||||
06711704
|
Restek™
RK 21650 |
0.25 in. Injection Port, 0.25 to 0.187 in. Column |
Each for $157.08
|
|
|||||
Description
- Use 0.125 in. and 0.19 in. OD columns in 1/4 in. on-column injection ports
- Slotted design prevents carrier gas occlusion
- Vespel/graphite reducing ferrules make installation easy
- Includes all nuts and ferrules used to attach tubing to the injector or detector
Specifications
| Adapter |