Learn More
Restek™ Open-End Wrench Sets
For tightening a wide variety of chromatography fittings
Supplier:  Restek™ RK 20387
Encompass
 View more versions of this product
Catalog No. 06710604
Only null left
Description
Description
Model 20387 Set:
  • 1/4 in. x 5/16 in.
  • 3/8 in. x 7/16 in.
  • 7/16 in. x 1/2 in.
  • 1/2 in. x 9/16 in.
Model 22334 Set (For use with Shimadzu 17A, 2010 and 2014 Capillary Installation Gauge):
  • 1/4 in. x 5/16 in.
  • 10mm x 11mm (0.39 in. x 0.43 in.)
Specifications
Chemical Identifiers
Open End Wrench Set
Specifications
Open End Wrench Set
Tightening Chromatography Fittings
Product Suggestions
Product Suggestions