missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Humidification Chamber DFS Item

Provides humidified air to remove volatile organic contaminants when cleaning SilcoCan or TO-Can canisters.

Supplier:  Restek™ RK24282

Catalog No. 06712280


Only null left
Explore available promotions

Description

Description

  • Made of acrylic and withstands pressure up to 90psi
  • Inlet (0.25 in.) and outlet compression fittings allow easy connection to pressure lines on cleaning system
  • Includes an easy-to-open lid for filling with water
Specifications

Specifications

Humidification Chamber
1 Each
90 psi
Remove Volatile Organic Contaminants When Cleaning SilcoCan or TO-Can Canisters
Acrylic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions