Learn More
Restek™ High-Temperature String
Use to restring capillary columns, attach column connectors to column cages, or hold the column in the GC oven.
$113.75 - $730.36
Specifications
Type High-Temperature String
View More Specs
Products 2
Catalog Number Mfr. No. Overall Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Overall Length (Metric) Price Quantity & Availability  
06710499
Encompass
View Documents Promotion Details
Restek™
RK 20109
10 m
Each for $113.75
Only null left
06710849
Encompass
View Documents Promotion Details
Restek™
RK 20618
450 m
Each for $730.36
Only null left
Description
Description
  • Capable of withstanding temperatures to 400°C
Specifications
Specifications
High-Temperature String