Learn More
Restek™ Ferrules for M4 Fittings
For QCQ Thermo™ Finnigan™ 8000 and Trace™ 2000 columns
Supplier:  Restek™ RK 20280
Encompass
Catalog No. 06717663
Only null left
Description
Description
  • 100% graphite
  • Made of high-purity materials that will not contribute background impurities for sensitive and selective detectors
  • Designed to seat securely in a variety of chromatography tube and pipe fittings
  • High-density and contain no binders that can off-gas or absorb analytes
  • With a smoother surface and cleaner edges that conventional graphite ferrules
  • Ferrules are very soft and can be easily deformed for an effective seal with fused silica or glass columns
  • Stable to 450°C (842°F)
Specifications
Specifications
Ferrule
2/Pk.
Graphite
M4 Fittings for QCQ Thermo Finnigan 8000 and TRACE 2000 Columns
450°C
2/Pk.
0.4 mm
Product Suggestions
Product Suggestions