Learn More
Restek™ Detector Lamps
Catalog No. 06712808
Encompass
Change view
Click to view available options
Type:
Deuterium Lamp
Lamp
For Use With (Equipment):
2487 Water Detector
486 Waters Detector
Agilent 8453 HPLC System
Agilent DAD G1315 A, G1365 A HPLC System
Agilent VWD G1314 A HPLC System
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
06712808 Lamp Agilent DAD G1315 A, G1365 A HPLC System
06712809 Lamp Agilent VWD G1314 A HPLC System
06712810 Deuterium Lamp Agilent 8453 HPLC System
06712919 Deuterium Lamp 2487 Water Detector
06712920 Deuterium Lamp 486 Waters Detector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06712808 Supplier Restek™ Supplier No. RK 25261
Used for waters detectors or for Agilent HPLC Systems
  • Standard lamps have nominal 1,000-hour life
Specifications
Type Lamp
For Use With (Equipment) Agilent DAD G1315 A, G1365 A HPLC System
Includes 1 Chromatography Detector Lamp