Learn More
Restek™ CGA Fittings
Catalog No. 06711530
Encompass
Change view
Click to view available options
Type:
CGA 350 Fitting
CGA 580 Fitting
CGA 590 Fitting
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06711530 CGA 590 Fitting
06711528 CGA 580 Fitting
06711529 CGA 350 Fitting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06711530 Supplier Restek™ Supplier No. RK 21338
Only null left
CGA-specified nuts and nipples with internal frit, 0.25 in. NPT nickel-plated brass.
  • Available in 3 in. or 3.5 in. lengths
Specifications
Type CGA 590 Fitting
Material Nickel-plated Brass