Learn More
Restek™ CarboPrep Cartridges
Description
- Improved recovery of sulfonylurea herbicides, phenols, carbamates, and triazine herbicides, compared to C18 and C8 cartridges
- Wide range of selectivity for both analytes and their metabolites or degradation products
- Rapid sampling flow rates; uncompromised recoveries
- Maximum capacity for contaminant cleanup
- Controlled manufacturing improves cleanliness, ensures reproducible performance
Specifications
Specifications
| Base Material | Silica |
| Bed Weight | 500 mg |
| For Use With (Application) | Sample Extraction of Organic Compounds and Extract Cleanup of Matrix Interferences |
| Sorbent | CarboPrep 90 |
| Type | SPE Cartridge |
| Volume (Metric) | 6 mL |
| Surface Area (Metric) | 90 m2/g |
| Tube Size | 6 mL, 500 mg |