Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC DFS Item
Fluted tip guides the capillary column into the jet
Supplier:  Restek™ RK21683
 View more versions of this product
Catalog No. 06711724
Only null left
Explore available promotions
Description
Description
  • Threads specially coated for easy installation and removal
  • Standard version undergoes special processing ensures the highest degree of cleanliness
  • High-performance Siltek™ version is extremely inert for use with active compounds
Specifications
Specifications
High-Performance Siltek Treated
3 Pack
Agilent 6890/6850
Product Suggestions
Product Suggestions