missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Blood Alcohol Mix Resolution Control Standard

Catalog No. 06802610
Encompass
Change view
Click to view available options
Quantity:
1 mL/Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06802610 1 mL/Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06802610 Supplier Restek™ Supplier No. RK 36256

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

To verify the retention time for each compound normally included in a blood alcohol test and to verify that the compounds are resolved from and do not interfere with one another.

  • Concentration in Solvent: 0.100g/dL each in water
  • Volume: 1mL/ampule
  • Shelf Life: 6 months
Compounds
  • Acetaldehyde (75-07-0)
  • Acetone (67-64-1)
  • Acetonitrile (75-05-8)
  • Ethanol (64-17-5)
  • Ethyl acetate (141-78-6)
  • 2-Propanol (Isopropanol) (67-63-0)
  • Methanol (67-56-1)
  • Methyl ethyl ketone (78-93-3)

Specifications

Concentration 0.1 g/dL
Type Blood Alcohol Mix Resolution Control Standard
Includes Acetaldehyde, Acetone, Acetonitrile, Ethanol, Ethyl Acetate, Isopropanol, Methanol, Methyl Ethyl Ketone
Intended Use Verify the retention time for each compound normally included in a Blood Alcohol test and to verify that the compounds are resolved from and do not interfere with one another
Quantity 1 mL/Ampule