Learn More
Restek™ 1/4in. Stainless-Steel Frits
Manufactured from fine, chromatographic grade 316 stainless steel.
Supplier: Restek™ RK 25072
Description
- Quantity: 10/Pk.
Chemical Identifiers
| Stainless Steel | |
| HPLC Frit |
Specifications
| Stainless Steel | |
| 4.6 mm | |
| HPLC Frit |
| 0.5 μm | |
| 0.18 in. |