missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Responder™ CSM ™ Suits

Catalog No. p-26259
Change view
Click to view available options
Size:
X-Large
Style:
Level A suit (gas tight)
Level B suit (nongas tight)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
190881219D X-Large Level B suit (nongas tight)
190881218D X-Large Level A suit (gas tight)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 190881219D Supplier DuPont™ Supplier No. RC128TTNXL000100
Only null left

Tested by U.S. Government-certified facility

Limited-use suits are designed specifically for dealing with deadly chemical-warfare agents, or Chemical Surety Materials (CSM). Tychem™ Responder CSM fabric is made to military specifications, with 100% inflation testing plus lot testing of suit construction and fabric.

  • Level A Fully Encapsulating (Gastight) Suit
    • Front-entry suit with gastight PVC zipper provides maximum protection for skin and eyes
    • Hood features a large 40mil PVC faceshield
    • Expanded back allows room for SCBA
    • Two exhaust valves reduce internal pressure buildup
    • Replaceable butyl gloves attached with ring-and-clamp system
    • Triple overlapping storm flap
    • Attached sock boots with boot flap
    • Double-sealed seams and 5mil PTFE faceshield overlay
    • Weight: 9 lb. (4.1kg)
    Level B Fully Encapsulating (Non-Gastight) Coverall
    • Zipper front with attached respirator hood
    • Butyl gloves attached with jam-fit glove system
    • Attached sock boots with splash guards
    • Double-sealed seams
    • Extended double-storm flap works with hood to accommodate respirator mask
    • Can be used with optional overhood to protect SCBA
    • Weight: 5 lb. (2.3kg)

    Specifications

    Color Tan
    For Use With (Application) Agriculture, general industry, construction and municipalities
    Material Tychem
    Size X-Large
    Ankle Style Attached socks, Boot overflaps
    Closure Type Zipper with hook-and-loop seal and double storm flap
    Cuff Style Attached gloves
    Description Standard width faceshield, 2 exhaust valves, attached Guardian™ IBA-35, 35mil Butyl gloves, socks with outer boot flaps, double storm flap with hook and loop closure; Non-woven Polyester construction
    Seam Type Taped
    Style Level B suit (nongas tight)
    Thickness 18 mil
    Show More Show Less