Learn More
Velp Scientifica™ RESPIROMETRIC Sensor Sys6
Catalog No. 07222013
Change view
Click to view available options
Voltage:
115 V
Catalog No. Voltage
07222013 115 V
1 options
Catalog No. 07222013 Supplier Velp Scientifica™ Supplier No. SA10210146
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The compact, reliable, cloud-enabled solution for BOD determination and aerobic respirometric tests.
  • Designed to provide a smart and cloud-enabled solution to laboratories of water and wastewater quality control
  • BOD (Biochemical Oxygen Demand) Determination is a simple and easy way for monitoring the microorganisms' activity in water samples with highly reliable results
  • In particular, microorganisms present or inoculated in a water sample containing biodegradable organic substance, consume oxygen for their metabolic activity and produce a corresponding volume of carbon dioxide
  • Carbon dioxide gets absorbed by a strong alkali positioned in the neck of the bottle
  • Result of this process is a decrease in gas pressure
  • BOD measures this change in gas pressure between the beginning to the end of the analysis and gives a result expressed in mg/L
Specifications
Type Sensor System
IP Rating IP54
Voltage 115 V
Certifications/Compliance CE