missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dentec Safety™ Respirator Wipes
Conveniently packaged wipes clean and disinfect respirators and other PPE equipment.
$161.60 - $161.60
Specifications
| Product Type | Respirator Wipes |
|---|---|
| For Use With (Equipment) | Facepieces |
| Quantity | 10 Boxes; 100/Box |
| Dimensions (L x W) | 5 x 8 in. (unfolded) |
Description
- Small packages can be stored anywhere - pocket, purse, lunch boxes, etc
- Safe and effective for cleaning respirators, hard hat suspensions, and other personal protective equipment
- Also useful for cleaning phones and keyboards
- Measures 5 x 8 inches when unfolded
Specifications
| Respirator Wipes | |
| Facepieces | |
| 10 Boxes; 100/Box | |
| 5 x 8 in. (unfolded) |