Learn More
United Scientific™ Resonance Box With Tuning Fork
Tuning forks are available in many different frequencies.
Supplier: United Scientific™ TFRB341
Description
- Wooden resonator box with tuning fork and mallet.
- Available in four different frequencies.
- Resonator box measures 18 cm x 9 cm x 5cm.
Chemical Identifiers
| Fork Resonance Apparatus |
Specifications
| Fork Resonance Apparatus | |
| 341.3 Hz |