missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ DNA Dilution Buffer

Catalog No. 4405587C
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4405587C 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4405587C Supplier Applied Biosystems™ Supplier No. 4405587C*

missing translation for 'validMainframeMsg'

DNA Dilution Buffer, a component of the ResDNASEQ Residual DNA Quantitation kits, is offered separately here for those kit users who need additional elution buffer.

DNA Dilution Buffer, a component of the ResDNASEQ Residual DNA Quantitation kits, is offered separately here for those kit users who need additional elution buffer.

As with other products in the Pharma Analytics Sample Preparation product line, DNA Dilution Buffer:

  • Is completely supported by our global manufacturing and distribution centers
  • Has a Certificate of Analysis available
  • Has a shelf-life of up to 12 months

Specifications

Chemical Name or Material Dilution Buffer
Physical Form Liquid
Quantity 7 mL

For Research Use Only. Not for use in diagnostic procedures.