Learn More
Restek™ Res-Sil B Packings DFS Item
Unique separation of saturated and unsaturated hydrocarbons.
$132.75 - $155.72
Specifications
Type Packing Material
View More Specs
Products 2
Catalog Number Mfr. No. Particle Size Material Price Quantity  
Catalog Number Mfr. No. Particle Size Material Price Quantity  
06712650 DFS Item
View Documents
Restek™
RK25080
80/100 Mesh ResSil B
Each for $132.75
Only null left
06712651 DFS Item
View Documents
Restek™
RK25081
80/100 Mesh 1% TCEP on ResSil B
Each for $155.72
Only null left
Description
Description
  • Innovative bonding chemistry for batch-to-batch reproducibility, excellent thermal stability, and long life
  • Wide range of bonded phases available
  • Equivalent to Durapak and Porasil packings
Specifications
Specifications
Packing Material
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'