Learn More
Danaher Corp Molecular Devices Microplate, 96-Well, Low Profile, Polystyrene
Catalog No. 5000014604
Encompass
Low-profile 96-well polystyrene microplate with round, flat-bottom wells supplied sterile with covers. The plate's low-profile format and optical flat bottom make it suitable for low-clearance microplate readers and automated workflows for absorbance, fluorescence, and cell-based assays.
  • Polystyrene construction for optical clarity and assay compatibility.
  • 96 round wells with flat bottoms for uniform optical measurements.
  • Low-profile format, approximately 10 mm high, for low-clearance readers.
  • Working volume of about 180 L per well.
  • Supplied sterile with covers to minimize contamination.
  • Packaged in sleeves of 10, 160 per case for bulk workflows.
  • Compatible with automated handlers and standard microplate readers.
Catalog No. 5000014604 Supplier Danaher Corp Supplier No. X6011
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Low-profile 96-well polystyrene microplate with round, flat-bottom wells supplied sterile with covers. The plate's low-profile format and optical flat bottom make it suitable for low-clearance microplate readers and automated workflows for absorbance, fluorescence, and cell-based assays.
  • Polystyrene construction for optical clarity and assay compatibility.
  • 96 round wells with flat bottoms for uniform optical measurements.
  • Low-profile format, approximately 10 mm high, for low-clearance readers.
  • Working volume of about 180 L per well.
  • Supplied sterile with covers to minimize contamination.
  • Packaged in sleeves of 10, 160 per case for bulk workflows.
  • Compatible with automated handlers and standard microplate readers.