Learn More
BRANDTECH™ BRAND™ Replacement Tip Cones and O-Rings for Transferpette™ S, Transferpette™ Electronic, and Transferpette™ Pro Multichannel Pipettes
Catalog No. 13688677
Change view
Click to view available options
For Use With (Equipment):
Electronic Pipette, 1 to 20 μL
Multichannel Pipette, 0.5 to 10 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13688677 Multichannel Pipette, 0.5 to 10 μL
13689221 Electronic Pipette, 1 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13688677 Supplier BRANDTECH™ BRAND™ Supplier No. 705677
Only null left
Spare parts for Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro, multichannel pipettes.
  • 705677: Includes two replacement tip cones for 0.5 to 10 μL nominal capacity Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro pipettes with dual O-ring design, complete with mounted O-rings
  • 705678: Includes two replacement tip cones for 1 to 20 μL nominal capacity Transferpette™ electronic pipettes with dual O-ring design, complete with mounted O-rings
  • 705680: Includes two replacement tip cones compatible with older models of BRAND Transferpette™ electronic 1 to 20 μL multichannel pipettes with a single O-ring design
  • 703380: Includes 24 O-rings; designed for use with Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro multichannel pipettes with nominal capacities of 0.5 to 10 μL and 1 to 20 μL
Specifications
For Use With Transferpette S, Transferpette Electronic, Transferpette Pro
Material Polypropylene
Type O-ring Kit
For Use With (Equipment) Multichannel Pipette, 0.5 to 10 μL
Includes Nose cones, O-rings, seals, instructions
Quantity 2/Pk.