Learn More
Synthware™ Replacement Straight Bore Stopcock
Supplied with PTFE plug, one PTFE washer and one O-ring.
Supplier: Synthware™ S181635
Specifications
| Straight Bore Stopcock | |
| Borosilicate Glass | |
| Straight Bore Tap | |
| PTFE |
| 6 mm | |
| Organic Synthesis | |
| PTFE plug, One PTFE washer, One o-ring |