Learn More
Allegro™ Fit Test Kit Accessory, Smoke Tube
Catalog No. 19083018
Change view
Click to view available options
:
Smoke Tubes 6/pk
Catalog No.
19083018 Smoke Tubes 6/pk
1 options
Catalog No. 19083018 Supplier Allegro™ Supplier No. 205001
Only null left
Allegro™ Fit Test Kit Accessory, Smoke Tube is used for qualitative fit testing of air purifying respirators.
  • Replacement tubes
  • For qualitative fit testing of air purifying respirators
Specifications
Product Type Smoke Tube
Certifications/Compliance OSHA
For Use With (Application) For qualitative fit testing of air purifying respirators
For Use With (Equipment) Allegro fit test kit
Model 2050-01
Quantity 6/Pk.