missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Replacement Sleeves for Clamps

Catalog No. 02216254
Change view
Click to view available options
Size:
Small
Medium
Large
Material:
Fiberglass
Vinyl
Color:
Black
Red
For Use With (Equipment):
2-prong Clamps
3-prong Multipurpose Clamps
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Color For Use With (Equipment)
02216254 Large Fiberglass Black 3-prong Multipurpose Clamps
02216252 Medium Fiberglass Black 3-prong Multipurpose Clamps
02216245 Medium Vinyl Red 2-prong Clamps
02216253 Large Vinyl Red 3-prong Multipurpose Clamps
02216248 Large Fiberglass Black 2-prong Clamps
02216250 Small Fiberglass Black 3-prong Multipurpose Clamps
02216247 Large Vinyl Red 2-prong Clamps
02216251 Medium Vinyl Red 3-prong Multipurpose Clamps
02216246 Medium Fiberglass Black 2-prong Clamps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02216254 Supplier Troemner™ Supplier No. 80862744

missing translation for 'validMainframeMsg'

Sleeves are easily removed for cleaning or replacement

  • Available in vinyl or fiberglass
  • Fiberglass sleeves are recommended for all applications above 100°C (212°F)

Specifications

Quantity 3/Pk.
Type Replacement Sleeve
Material Fiberglass
For Use With (Equipment) 3-prong Multipurpose Clamps
Length (Metric) 7 cm
Size Large
Color Black