Learn More
Troemner™ Replacement Sleeves for Clamps
Sleeves are easily removed for cleaning or replacement
 View more versions of this product
Catalog No. 02216248
Only null left
Description
Description
  • Available in vinyl or fiberglass
  • Fiberglass sleeves are recommended for all applications above 100°C (212°F)
Specifications
Chemical Identifiers
Replacement Sleeve
Specifications
Replacement Sleeve
2-prong Clamps
Black
Fiberglass
Large
Product Suggestions
Product Suggestions