Learn More
Thermo Scientific™ Replacement Sealing O-Rings for HIGHConic™ II Fixed-Angle Rotors
Description
Replace cracked, dry or otherwise faulty O-rings with Thermo Scientific™ Replacement Sealing O-Rings, a replacement set of two that includes grease.
Specifications
Specifications
| For Use With (Equipment) | HiGhConic II Fixed-angle Rotor |
| Description | Replacement Sealing O-rings |
| Product Type | O-ring |