Learn More
Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator
Supplier:  Mettler Toledo™ 23936
 View more versions of this product
Catalog No. 01910747
Only null left
Description
Description
Coulometric Cells
  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
Specifications
Specifications
Solvent Drain/Delivery Tube
PTFE
Product Suggestions
Product Suggestions