missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator

Catalog No. 01910738
Change view
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Coulometric Reagent Set
Drying Tube
Septa
Solvent Drain/Delivery Tube
Titration Vessel
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01910738 Coulometric Cell without Diaphragm
01910737 Coulometric Cell with Diaphragm
01911144 Coulometric Reagent Set
01910735 Titration Vessel
01910744 Septa
01910747 Solvent Drain/Delivery Tube
01910741 Drying Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01910738 Supplier Mettler Toledo™ Supplier No. 51108753

Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator

Coulometric Cells

  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents

Specifications

Type Coulometric Cell without Diaphragm
For Use With (Application) General coulometric KF titration
Material Glass
For Use With (Equipment) C20 Coulometric Titrator