Learn More
Analox Sensor Technology O2NE+, REPL SNSR, MFR <11/2016
Supplier:  Analox Sensor Technology 91001021
Encompass
Analox O2NE+ Replacement Oxygen Sensor for models manufactured before November 2016. Calibration is required after sensor replacement.
Catalog No. FSTP9779162
Only null left