Learn More
Thermo Scientific™ Replacement Microcentrifuge Rotor Lids
Description
Combine Thermo Scientific™ Replacement Microcentrifuge Rotor Lids with the microcentrifuge rotor.
ClickSeal™ Biocontainment Lid:
- For use with 24 × 1.5/2.0 mL Rotor (75003424), PCR 4 × 8 Rotor (75003440)
- Unique design clicks when proper seal is made, providing assurance that your samples are securely contained, as well as shortens retrieval time with one click opening
- With PPSU rotor lid, view contents of rotor for added safety and convenience
Screw-on Lid:
- For use with 36 × 0.5 mL Rotor (75003436), Dual Row 18 × 2.00.5 mL Rotor (75003418), PCR 8 × 8 Rotor (75003489)
Ordering Information:
Sold individually.
Specifications
Specifications
| For Use With (Equipment) | 24 x 1.5/2 mL Rotor (75003424), PCR 4 x 8 Rotor |
| Description | ClickSeal™ Biocontainment Lid |
| Product Type | ClickSeal Lid |