Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
Catalog No. 75005730
Change view
Click to view available options
Product Type:
ClickSeal Biocontainment Lid
O-ring
For Use With (Equipment):
MicroClick 24 x 2 Rotor
MicroClick 24 x 2 Rotor Lid
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
75005730 ClickSeal Biocontainment Lid MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
75003405 O-ring MicroClick 24 x 2 Rotor Lid
75005725 ClickSeal Biocontainment Lid MicroClick 24 x 2 Rotor
75005726 O-ring MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 75005730 Supplier Thermo Scientific™ Supplier No. 75005730
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
Specifications
For Use With (Equipment) MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Product Type ClickSeal Biocontainment Lid