Learn More
Thermo Scientific™ Replacement Seals for Thermo Scientific™ MicroClick and 8 x 8 PCR Strip Rotors
A range of options for secure closure on microcentrifuge rotors to help contain samples.
Supplier:  Thermo Scientific™ 75005726
 View more versions of this product
Catalog No. 75005726
Only null left
Description
Description
A range of options for secure closure on microcentrifuge rotors to help contain samples.
  • O-rings for inner and outer seals
  • ClickSeal lids for optimal biocontainment
  • Available for a variety of sizes and models
Specifications
Specifications
O-ring
MicroClick 30 x 2 and 8 x 8 PCR Strip Rotors
Product Suggestions
Product Suggestions